Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSTPIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024430.3 | NP_077748.3 | 15 | 1985 | ||
XM_011526253.2 | XP_011524555.1 | 14 | 1950 | |||
XM_011526254.1 | XP_011524556.1 | 14 | 1862 | |||
XM_011526256.1 | XP_011524558.1 | 14 | 1861 | |||
GenBank mRNA | AK023100.1 | - | 11 | 1553 | ||
BC026223.1 | - | 1 | 724 | |||
BC035395.1 | - | 14 | 1861 | 4524856 |