Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDADC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193478.1 | NP_001180407.1 | 10 | 1924 | ||
NM_030911.3 | NP_112173.1 | 10 | 3018 | |||
NR_036439.1 | 9 | 2840 | ||||
XM_006719871.3 | XP_006719934.1 | 9 | 2787 | |||
XM_011535250.2 | XP_011533552.1 | 10 | 2982 | |||
XM_011535251.1 | XP_011533553.1 | 10 | 2962 | |||
XM_011535253.1 | XP_011533555.1 | 10 | 2914 | |||
XM_017020778.1 | XP_016876267.1 | 9 | 2721 | |||
XM_017020779.1 | XP_016876268.1 | 11 | 3138 | |||
XM_017020780.1 | XP_016876269.1 | 11 | 3485 | |||
XM_017020781.1 | XP_016876270.1 | 10 | 2957 | |||
XM_017020782.1 | XP_016876271.1 | 11 | 3061 | |||
XM_017020783.1 | XP_016876272.1 | 9 | 3182 | |||
XM_017020784.1 | XP_016876273.1 | 8 | 2753 | |||
GenBank mRNA | AL137554.1 | - | 4 | 1694 | ||
AV648588.1 | - | 1 | 119 | |||
AY027525.1 | - | 10 | 2967 | |||
BC048092.1 | - | 11 | 2985 | |||
BC125202.1 | - | 10 | 2932 | 40123430 |