Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076785.2 | NP_001070253.1 | 4 | 390 | ||
NM_003983.5 | NP_003974.3 | 3 | 303 | |||
XM_011523433.1 | XP_011521735.1 | 3 | 405 | |||
XM_011523434.1 | XP_011521736.1 | 3 | 470 | |||
XM_011523438.1 | XP_011521740.1 | 2 | 860 | |||
XM_011523439.1 | XP_011521741.1 | 3 | 989 | |||
GenBank mRNA | AK310866.1 | - | 3 | 269 | ||
BC028216.1 | - | 3 | 259 | 5214340 | ||
BC113100.1 | - | 3 | 290 | |||
CR749291.1 | - | 3 | 289 | |||
CR749475.1 | - | 3 | 272 | |||
D87432.1 | - | 3 | 338 | |||
DB107640.1 | - | 3 | 303 |