Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160706.1 | NP_001154178.1 | 10 | 752 | ||
NM_003843.3 | NP_003834.3 | 11 | 818 | |||
NM_144777.2 | NP_659001.2 | 11 | 818 | |||
XM_005266578.1 | XP_005266635.1 | 10 | 729 | |||
XM_006719882.1 | XP_006719945.1 | 11 | 764 | |||
XM_006719884.1 | XP_006719947.1 | 10 | 698 | |||
XM_006719885.1 | XP_006719948.1 | 11 | 764 | |||
XM_011535281.1 | XP_011533583.1 | 11 | 764 | |||
XM_011535282.1 | XP_011533584.1 | 11 | 764 | |||
XM_011535283.1 | XP_011533585.1 | 11 | 764 | |||
XM_011535284.1 | XP_011533586.1 | 11 | 764 | |||
XM_011535285.1 | XP_011533587.1 | 10 | 774 | |||
XM_011535286.1 | XP_011533588.1 | 11 | 764 | |||
XM_011535287.1 | XP_011533589.1 | 11 | 764 | |||
XM_011535288.1 | XP_011533590.1 | 9 | 628 | |||
XM_011535289.1 | XP_011533591.1 | 11 | 764 | |||
XM_011535290.1 | XP_011533592.1 | 11 | 764 | |||
XM_011535291.1 | XP_011533593.1 | 11 | 764 | |||
XM_017020805.1 | XP_016876294.1 | 11 | 764 | |||
GenBank mRNA | AF045941.1 | - | 11 | 734 | ||
AK025320.1 | - | 11 | 732 | |||
AK222720.1 | - | 11 | 732 | |||
AK301659.1 | - | 10 | 666 | |||
BC020726.1 | - | 10 | 699 | |||
BC047536.1 | - | 11 | 893 | 5271857 |