Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP4K4 SIRNA Assays ›
Gene Symbol:
MAP4K4
Gene Name:
mitogen-activated protein kinase kinase kinase kinase 4
Gene Aliases:
FLH21957, HEL-S-31, HGK, MEKKK4, NIK
Chromosome Location:
Chr.2: 101697703 - 101894690 on Build GRCh38
UniGene ID:
Hs.701013
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138645
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP4K4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242559.1
NP_001229488.1
13
1268
NM_001242560.1
NP_001229489.1
13
1641
NM_004834.4
NP_004825.3
13
1641
NM_145686.3
NP_663719.2
13
1641
NM_145687.3
NP_663720.1
13
1268
XM_005264048.2
XP_005264105.1
13
1337
XM_005264050.3
XP_005264107.1
13
1337
XM_005264052.3
XP_005264109.1
13
1337
XM_005264057.3
XP_005264114.1
13
1337
XM_005264058.3
XP_005264115.1
13
1337
XM_005264059.3
XP_005264116.1
13
1337
XM_005264060.3
XP_005264117.1
13
1337
XM_005264062.3
XP_005264119.1
13
1337
XM_005264063.3
XP_005264120.1
13
1337
XM_005264064.3
XP_005264121.1
13
1337
XM_005264065.3
XP_005264122.1
13
1337
XM_005264066.3
XP_005264123.1
13
1337
XM_005264068.3
XP_005264125.1
13
1337
XM_005264069.2
XP_005264126.1
13
1337
XM_005264071.2
XP_005264128.1
13
1337
XM_006712865.2
XP_006712928.1
13
1337
XM_006712868.2
XP_006712931.1
13
1337
XM_006712869.2
XP_006712932.1
13
1337
XM_017005349.1
XP_016860838.1
13
1337
XM_017005350.1
XP_016860839.1
13
1337
XM_017005351.1
XP_016860840.1
13
1337
XM_017005352.1
XP_016860841.1
13
1337
XM_017005353.1
XP_016860842.1
13
1337
XM_017005354.1
XP_016860843.1
13
1337
XM_017005355.1
XP_016860844.1
13
1337
XM_017005356.1
XP_016860845.1
13
1337
XM_017005357.1
XP_016860846.1
13
1337
XM_017005358.1
XP_016860847.1
13
1337
XM_017005359.1
XP_016860848.1
13
1337
XM_017005360.1
XP_016860849.1
13
1337
XM_017005361.1
XP_016860850.1
13
1337
XM_017005362.1
XP_016860851.1
13
1337
XM_017005363.1
XP_016860852.1
13
1337
XM_017005364.1
XP_016860853.1
13
1337
XM_017005365.1
XP_016860854.1
13
1337
XM_017005366.1
XP_016860855.1
13
1337
XM_017005367.1
XP_016860856.1
13
1337
XM_017005368.1
XP_016860857.1
13
1337
XM_017005369.1
XP_016860858.1
13
1337
XM_017005370.1
XP_016860859.1
13
1337
XM_017005371.1
XP_016860860.1
13
1337
XM_017005372.1
XP_016860861.1
13
1337
XM_017005373.1
XP_016860862.1
13
1337
XM_017005374.1
XP_016860863.1
13
1337
XM_017005375.1
XP_016860864.1
13
1337
GenBank mRNA
AB014587.1
-
11
1152
AK025610.1
-
2
106
AK074900.1
-
9
874
AK295610.1
-
11
1233
AK296396.1
-
13
1337
AK307879.1
-
9
823
AL137755.1
-
4
481
AY212247.1
-
13
1263
AY834276.1
-
9
822
EU794634.1
-
13
1318
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction by protein phosphorylation
microvillus assembly
intracellular signal transduction
negative regulation of apoptotic process
regulation of JNK cascade
creatine kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0