Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USH1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297764.1 | NP_001284693.1 | 20 | 1981 | ||
NM_005709.3 | NP_005700.2 | 21 | 2038 | |||
NM_153676.3 | NP_710142.1 | 27 | 3047 | |||
NR_123738.1 | 20 | 1964 | ||||
XM_011519832.2 | XP_011518134.1 | 22 | 2191 | |||
GenBank mRNA | AB006955.1 | - | 20 | 2081 | ||
AB018687.1 | - | 21 | 2018 | |||
AF039699.1 | - | 20 | 1953 | |||
AF039700.1 | - | 21 | 2027 | |||
AK225614.1 | - | 21 | 2038 | |||
AK300936.1 | - | 21 | 1797 | |||
BC016057.1 | - | 20 | 1965 | 4867369 | ||
BX641129.1 | - | 1 | 1000 |