Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003818.3 | NP_003809.1 | 7, 8 | 1001 | ||
XM_006723660.1 | XP_006723723.1 | 7, 8 | 992 | |||
XM_011529390.1 | XP_011527692.1 | 8, 9 | 1324 | |||
GenBank mRNA | AF069532.1 | - | 6, 7 | 597 | ||
AK001414.1 | - | 7, 8 | 722 | |||
AK027695.1 | - | 7, 8 | 708 | |||
AK091045.1 | - | 8, 9 | 1324 | |||
AK303426.1 | - | 3, 4 | 275 | |||
AK304763.1 | - | 4, 5 | 465 | |||
AK315489.1 | - | 7, 8 | 982 | |||
BC025751.1 | - | 7, 8 | 722 | 5199748 | ||
Y16521.1 | - | 7, 8 | 927 |