Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPHK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142601.1 | NP_001136073.1 | 6 | 1329 | ||
NM_001142602.1 | NP_001136074.1 | 5 | 1374 | |||
NM_021972.3 | NP_068807.2 | 6 | 1371 | |||
NM_182965.2 | NP_892010.2 | 6 | 1692 | |||
XM_005257766.2 | XP_005257823.1 | 6 | 1684 | |||
GenBank mRNA | AF200328.1 | - | 5 | 955 | ||
AF238083.1 | - | 6 | 1273 | |||
AF266756.1 | - | 5 | 1206 | |||
AJ245504.1 | - | 4 | 928 | |||
AK022402.1 | - | 6 | 1329 | |||
AK023393.1 | - | 5 | 1286 | |||
AK095578.1 | - | 5 | 2014 | |||
AK122781.1 | - | 8 | 1746 | |||
AK225250.1 | - | 6 | 1292 | |||
AK225258.1 | - | 6 | 1158 | |||
AK292294.1 | - | 6 | 1416 | |||
AK292614.1 | - | 5 | 1290 | |||
BC004112.2 | - | 2 | 646 | |||
BC008040.2 | - | 6 | 1266 | 3831657 | ||
BC009419.1 | - | 6 | 1359 | 3506924 | ||
BC014439.1 | - | 6 | 1629 | 4871343 | ||
BC030553.1 | - | 6 | 1651 | 5213270 |