Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287489.1 | NP_001274418.1 | 44 | 5829 | ||
NM_004802.3 | NP_004793.2 | 27 | 3627 | |||
NM_194248.2 | NP_919224.1 | 44 | 5829 | |||
NM_194322.2 | NP_919303.1 | 26 | 3781 | |||
NM_194323.2 | NP_919304.1 | 27 | 3627 | |||
XM_017005338.1 | XP_016860827.1 | 44 | 5642 | |||
GenBank mRNA | AF107403.1 | - | 27 | 3627 | ||
AF183185.1 | - | 44 | 5829 | |||
AF183186.1 | - | 26 | 3781 | |||
AF183187.1 | - | 27 | 3627 | |||
AK296272.1 | - | 17 | 2184 |