Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000361.2 | NP_000352.1 | 1 | 2726 | ||
GenBank mRNA | AK123557.1 | - | 2 | 2540 | ||
BC035602.1 | - | 1 | 2718 | 5176531 | ||
M16552.1 | - | 1 | 2712 | |||
X05495.1 | - | 1 | 2716 |