Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000551.3 | NP_000542.1 | 3 | 830 | ||
NM_198156.2 | NP_937799.1 | 2 | 707 | |||
GenBank mRNA | AK304002.1 | - | 3 | 415 | ||
AK309560.1 | - | 3 | 754 | |||
AK315799.1 | - | 3 | 689 | |||
BC058831.1 | - | 2 | 558 | 6457060 | ||
KU178386.1 | - | 2 | 494 | |||
KU178387.1 | - | 3 | 617 | |||
L15409.1 | - | 3 | 830 |