Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000557.4 | NP_000548.2 | 2 | 2276 | ||
NM_001319138.1 | NP_001306067.1 | 4 | 2518 | |||
XM_011529075.2 | XP_011527377.1 | 4 | 2408 | |||
GenBank mRNA | AB593125.1 | - | 2 | 2264 | ||
BC032495.1 | - | 4 | 2518 | 5243109 | ||
U13660.1 | - | 2 | 2272 |