Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000660.5 | NP_000651.3 | 1, 2 | 1244 | ||
XM_011527242.1 | XP_011525544.1 | 1, 2 | 1237 | |||
GenBank mRNA | AK291907.1 | - | 1, 2 | 733 | ||
AK307742.1 | - | 1, 2 | 929 | |||
BC000125.1 | - | 1, 2 | 801 | 3351664 | ||
BC001180.1 | - | 1, 2 | 801 | 3356605 | ||
BC022242.1 | - | 1, 2 | 724 | 4399762 | ||
BT007245.1 | - | 1, 2 | 355 | |||
X02812.1 | - | 1, 2 | 1196 |