Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001063.3 | NP_001054.1 | 2 | 500 | ||
XM_017007089.1 | XP_016862578.1 | 8 | 1850 | |||
XM_017007090.1 | XP_016862579.1 | 8 | 2843 | |||
GenBank mRNA | AJ252279.1 | - | 2 | 351 | ||
AK222755.1 | - | 2 | 240 | |||
AK293842.1 | - | 2 | 240 | |||
AK295334.1 | - | 2 | 240 | |||
AK303753.1 | - | 2 | 240 | |||
BC059367.1 | - | 2 | 238 | 5264299 | ||
CR936810.1 | - | 7 | 1921 | |||
DC352309.1 | - | 2 | 500 | |||
DQ923758.1 | - | 2 | 231 | |||
GQ472199.1 | - | 2 | 238 | |||
M12530.1 | - | 2 | 222 | |||
S95936.1 | - | 2 | 271 |