Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001063.3 | NP_001054.1 | 17 | 2408 | ||
XM_017007089.1 | XP_016862578.1 | 23 | 3758 | |||
XM_017007090.1 | XP_016862579.1 | 23 | 4751 | |||
GenBank mRNA | AK126941.1 | - | 2 | 4447 | ||
AK222755.1 | - | 17 | 2148 | |||
AK295334.1 | - | 17 | 2065 | |||
AK295419.1 | - | 16 | 1975 | |||
AK303753.1 | - | 17 | 2088 | |||
BC059367.1 | - | 17 | 2146 | 5264299 | ||
BX537660.1 | - | 2 | 2789 | |||
BX648533.1 | - | 17 | 1925 | |||
CR936810.1 | - | 22 | 3830 | |||
DQ923758.1 | - | 17 | 2139 | |||
GQ472199.1 | - | 17 | 2146 | |||
M12525.1 | - | 8 | 838 | |||
M12530.1 | - | 17 | 2130 | |||
S95936.1 | - | 17 | 2179 |