Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001074.2 | NP_001065.2 | 6 | 1672 | ||
XM_005265702.3 | XP_005265759.1 | 7 | 1376 | |||
XM_011532230.1 | XP_011530532.1 | 5 | 1453 | |||
GenBank mRNA | AK223142.1 | - | 6 | 1646 | ||
BC030974.1 | - | 6 | 1672 | 4593426 | ||
J05428.1 | - | 6 | 1640 |