Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078.3 | NP_001069.1 | 3 | 622 | ||
NM_001199834.1 | NP_001186763.1 | 3 | 436 | |||
NM_080682.2 | NP_542413.1 | 3 | 622 | |||
GenBank mRNA | AK223266.1 | - | 3 | 521 | ||
AK291732.1 | - | 3 | 520 | |||
AK296692.1 | - | 3 | 334 | |||
AU121762.1 | - | 3 | 522 | |||
BC017276.2 | - | 3 | 502 | 4845424 | ||
BC068490.1 | - | 3 | 520 | 30339170 | ||
BC085003.1 | - | 3 | 502 | 6375943 | ||
M30257.1 | - | 3 | 507 | |||
M60335.1 | - | 3 | 500 | |||
X53051.1 | - | 3 | 401 |