Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277961.1 | NP_001264890.1 | 1, 2 | 712 | ||
NM_001277962.1 | NP_001264891.1 | 1, 2 | 712 | |||
NM_003156.3 | NP_003147.2 | 1, 2 | 712 | |||
GenBank mRNA | AK314928.1 | - | 1, 2 | 233 | ||
BC021300.2 | - | 1, 2 | 704 | 4899542 | ||
HQ317451.1 | - | 1, 2 | 711 | |||
JX014264.1 | - | 1, 2 | 144 | |||
U52426.1 | - | 1, 2 | 709 |