Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031464.4 | NP_113652.2 | 7, 8 | 1763 | ||
XM_011537206.2 | XP_011535508.1 | 9, 10 | 2039 | |||
XM_011537207.2 | XP_011535509.1 | 8, 9 | 1497 | |||
XM_011537211.2 | XP_011535513.1 | 7, 8 | 1930 | |||
XM_011537213.2 | XP_011535515.1 | 6, 7 | 1882 | |||
XM_017021680.1 | XP_016877169.1 | 8, 9 | 1459 | |||
XM_017021681.1 | XP_016877170.1 | 7, 8 | 1411 | |||
XM_017021682.1 | XP_016877171.1 | 7, 8 | 1401 | |||
XM_017021684.1 | XP_016877173.1 | 8, 9 | 1459 | |||
XM_017021685.1 | XP_016877174.1 | 8, 9 | 1459 | |||
GenBank mRNA | AK299846.1 | - | 7, 8 | 1529 | ||
BC004540.2 | - | 6, 7 | 1402 | 3945902 |