Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282541.1 | NP_001269470.1 | 14 | 1362 | ||
NM_001282542.1 | NP_001269471.1 | 14 | 1362 | |||
NM_003176.3 | NP_003167.2 | 14 | 1359 | |||
NR_104211.1 | 14 | 1359 | ||||
XM_005271155.3 | XP_005271212.1 | 13 | 1683 | |||
XM_005271156.1 | XP_005271213.1 | 13 | 1157 | |||
XM_006710859.1 | XP_006710922.1 | 14 | 1270 | |||
XM_011542037.2 | XP_011540339.1 | 14 | 1326 | |||
XM_011542038.2 | XP_011540340.1 | 7 | 553 | |||
XM_011542039.1 | XP_011540341.1 | 14 | 1326 | |||
XM_017002184.1 | XP_016857673.1 | 14 | 1162 | |||
XM_017002185.1 | XP_016857674.1 | 8 | 588 | |||
GenBank mRNA | AF273043.1 | - | 14 | 1297 | ||
AK315268.1 | - | 14 | 1321 | |||
BC126266.1 | - | 14 | 1236 | 8991982 | ||
BC144019.1 | - | 14 | 1239 | 9052539 | ||
D67035.1 | - | 14 | 1295 | |||
X95654.1 | - | 14 | 1213 |