Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099691.2 | NP_001093161.1 | 6 | 946 | ||
NM_001308158.1 | NP_001295087.1 | 6 | 1116 | |||
NM_001308159.1 | NP_001295088.1 | 6 | 1113 | |||
NM_003236.3 | NP_003227.1 | 6 | 949 | |||
GenBank mRNA | BC005308.1 | - | 6 | 757 | 3828239 | |
DA229819.1 | - | 1 | 64 | |||
K03222.1 | - | 6 | 735 | |||
M31172.1 | - | 6 | 735 | |||
X70340.1 | - | 6 | 732 |