Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003855.3 | NP_003846.1 | 3 | 366 | ||
XM_005264039.3 | XP_005264096.1 | 2 | 248 | |||
XM_005264040.3 | XP_005264097.1 | 2 | 248 | |||
XM_011512099.1 | XP_011510401.1 | 2 | 248 | |||
XM_017005181.1 | XP_016860670.1 | 2 | 248 | |||
GenBank mRNA | AY192162.1 | - | 2 | 220 | ||
BC069575.1 | - | 2 | 244 | 7262302 | ||
BC093975.1 | - | 2 | 244 | 7939820 | ||
BC093977.1 | - | 2 | 244 | 7939822 | ||
BC143403.1 | - | 2 | 244 | 9051910 | ||
U43672.1 | - | 2 | 244 |