Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003247.3 | NP_003238.2 | 23 | 4974 | ||
GenBank mRNA | BC146676.1 | - | 22 | 4781 | 40148868 | |
BC150175.1 | - | 22 | 4785 | 40147958 | ||
BX641023.1 | - | 9 | 2429 | |||
L12350.1 | - | 23 | 4893 |