Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003262.3 | NP_003253.1 | 8 | 1082 | ||
XM_011513114.2 | XP_011511416.1 | 9 | 1159 | |||
GenBank mRNA | AK294504.1 | - | 3 | 246 | ||
BC012035.1 | - | 8 | 1066 | 4706586 | ||
BX648539.1 | - | 8 | 1066 | |||
D87127.1 | - | 8 | 1063 | |||
U93239.1 | - | 8 | 1063 |