Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003262.3 | NP_003253.1 | 8 | 1953 | ||
XM_011513114.2 | XP_011511416.1 | 9 | 2030 | |||
GenBank mRNA | BC012035.1 | - | 9 | 1384 | 4706586 | |
BX648539.1 | - | 8 | 1937 | |||
D87127.1 | - | 8 | 1934 | |||
U93239.1 | - | 8 | 1934 |