Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286102.1 | NP_001273031.1 | 9 | 1610 | ||
NM_003269.4 | NP_003260.1 | 9 | 2203 | |||
GenBank mRNA | AF220532.1 | - | 9 | 2202 | ||
AK131541.1 | - | 9 | 1610 | |||
BC028031.1 | - | 9 | 1700 | 5242079 |