Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003292.2 | NP_003283.2 | 1, 2 | 446 | ||
GenBank mRNA | AK309473.1 | - | 2, 3 | 481 | ||
BC016514.1 | - | 1, 2 | 320 | |||
BC094764.1 | - | 1, 2 | 375 | |||
U69668.1 | - | 1, 2 | 237 | |||
X63105.1 | - | 1, 2 | 446 | |||
X66397.1 | - | 1, 2 | 446 | |||
Y00672.1 | - | 1, 2 | 261 |