Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032287.2 | NP_001027458.1 | 3 | 414 | ||
NM_001127362.1 | NP_001120834.1 | 3 | 414 | |||
NM_003297.3 | NP_003288.2 | 3 | 414 | |||
XM_005269133.2 | XP_005269190.1 | 3 | 401 | |||
XM_006719584.2 | XP_006719647.1 | 3 | 401 | |||
XM_006719585.2 | XP_006719648.1 | 4 | 439 | |||
XM_011538716.2 | XP_011537018.1 | 3 | 401 | |||
XM_011538717.2 | XP_011537019.1 | 3 | 401 | |||
GenBank mRNA | AB307707.1 | - | 2 | 158 | ||
AK291319.1 | - | 3 | 378 | |||
BC026074.1 | - | 3 | 360 | |||
BC040141.1 | - | 3 | 366 | 5183723 | ||
DB266298.1 | - | 3 | 414 | |||
JQ308177.1 | - | 2 | 158 | |||
JQ308178.1 | - | 2 | 158 | |||
M21985.1 | - | 3 | 284 | |||
M29959.1 | - | 3 | 284 | |||
M29960.1 | - | 3 | 214 |