Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144766.2 | NP_001138238.1 | 4 | 421 | ||
NM_003315.3 | NP_003306.3 | 4 | 653 | |||
NR_029431.1 | 3 | 564 | ||||
XM_011525167.2 | XP_011523469.1 | 5 | 583 | |||
XM_011525168.2 | XP_011523470.1 | 5 | 543 | |||
XM_011525169.2 | XP_011523471.1 | 4 | 395 | |||
XM_017024994.1 | XP_016880483.1 | 5 | 577 | |||
GenBank mRNA | AB209834.1 | - | 4 | 339 | ||
AK291047.1 | - | 4 | 354 | |||
AK296602.1 | - | 4 | 375 | |||
AK298860.1 | - | 4 | 369 | |||
AK308016.1 | - | 4 | 407 | |||
AK316081.1 | - | 3 | 290 | |||
BC003601.1 | - | 3 | 276 | 3543074 | ||
BC011837.2 | - | 4 | 372 | 3945721 | ||
BC033772.1 | - | 4 | 398 | 3924081 | ||
BP378441.1 | - | 4 | 402 | |||
BX647209.1 | - | 4 | 609 | |||
U46571.1 | - | 4 | 368 |