Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003321.4 | NP_003312.3 | 2, 3 | 382 | ||
XM_011545928.2 | XP_011544230.1 | 2, 3 | 476 | |||
XM_017023619.1 | XP_016879108.1 | 3, 4 | 639 | |||
GenBank mRNA | BC001633.1 | - | 2, 3 | 288 | 3536952 | |
BC010041.2 | - | 2, 3 | 296 | 3629164 | ||
L38995.1 | - | 2, 3 | 241 | |||
S75463.1 | - | 2, 3 | 334 | |||
X84694.1 | - | 2, 3 | 241 |