Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003321.4 | NP_003312.3 | 10 | 1365 | ||
XM_011545928.2 | XP_011544230.1 | 10 | 1375 | |||
XM_017023619.1 | XP_016879108.1 | 11 | 1622 | |||
GenBank mRNA | BC001633.1 | - | 10 | 1271 | 3536952 | |
BC010041.2 | - | 10 | 1279 | 3629164 | ||
L38995.1 | - | 10 | 1224 | |||
S75463.1 | - | 10 | 1308 | |||
X84694.1 | - | 10 | 1224 |