Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003328.2 | NP_003319.2 | 9, 10 | 863 | ||
XM_011513747.2 | XP_011512049.1 | 8, 9 | 815 | |||
XM_011513748.2 | XP_011512050.1 | 2, 3 | 151 | |||
XM_017008580.1 | XP_016864069.1 | 10, 11 | 2065 | |||
XM_017008581.1 | XP_016864070.1 | 8, 9 | 1874 | |||
XM_017008582.1 | XP_016864071.1 | 3, 4 | 298 | |||
GenBank mRNA | L27071.1 | - | 9, 10 | 863 |