Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003331.4 | NP_003322.3 | 6 | 917 | ||
XM_011528245.1 | XP_011526547.1 | 6 | 827 | |||
XM_011528246.2 | XP_011526548.1 | 5 | 652 | |||
XM_011528247.1 | XP_011526549.1 | 4 | 429 | |||
XM_011528252.1 | XP_011526554.1 | 5 | 696 | |||
GenBank mRNA | AK222672.1 | - | 6 | 870 | ||
AK308360.1 | - | 2 | 132 | |||
BC014243.2 | - | 6 | 880 | 4591726 | ||
X54637.1 | - | 6 | 845 |