Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003365.2 | NP_003356.2 | 13 | 1552 | ||
GenBank mRNA | AA284910.1 | - | 1 | 42 | ||
AK129903.1 | - | 13 | 1532 | |||
AK300694.1 | - | 13 | 1487 | |||
BC009586.1 | - | 13 | 1523 | 4096628 | ||
D26485.1 | - | 13 | 1514 | |||
L16842.1 | - | 13 | 1924 |