Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVRAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003369.3 | NP_003360.2 | 15 | 2417 | ||
XM_011545226.2 | XP_011543528.1 | 16 | 2810 | |||
XM_017018224.1 | XP_016873713.1 | 14 | 2249 | |||
XM_017018225.1 | XP_016873714.1 | 13 | 2171 | |||
XM_017018227.1 | XP_016873716.1 | 11 | 1836 | |||
GenBank mRNA | AB012958.1 | - | 15 | 2354 | ||
AK095352.1 | - | 9 | 1994 | |||
AK222772.1 | - | 15 | 2308 | |||
AK291011.1 | - | 15 | 2395 | |||
AK293782.1 | - | 15 | 2158 | |||
AK296871.1 | - | 9 | 1537 | |||
AK299978.1 | - | 3 | 1154 | |||
AK316133.1 | - | 9 | 1604 | |||
BC064837.1 | - | 15 | 2327 | 6186851 |