Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003374.2 | NP_003365.1 | 9 | 1385 | ||
NR_036624.1 | 9 | 1463 | ||||
NR_036625.1 | 9 | 1466 | ||||
XM_005272075.3 | XP_005272132.1 | 9 | 1382 | |||
XM_017009821.1 | XP_016865310.1 | 9 | 1313 | |||
XM_017009822.1 | XP_016865311.1 | 9 | 1292 | |||
XM_017009823.1 | XP_016865312.1 | 9 | 1315 | |||
GenBank mRNA | AK095989.1 | - | 9 | 1385 | ||
AK122953.1 | - | 12 | 2788 | |||
AK293639.1 | - | 7 | 842 | |||
BC008482.1 | - | 9 | 1170 | 4332199 | ||
BC071168.1 | - | 9 | 1202 | 6023095 | ||
BC090042.1 | - | 9 | 1195 | 30395350 | ||
L06132.1 | - | 9 | 1239 |