Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EZR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111077.1 | NP_001104547.1 | 14 | 1854 | ||
NM_003379.4 | NP_003370.2 | 13 | 1888 | |||
XM_011536110.1 | XP_011534412.1 | 10 | 1265 | |||
GenBank mRNA | AK129727.1 | - | 7 | 810 | ||
AK296738.1 | - | 14 | 1683 | |||
AK299456.1 | - | 12 | 1717 | |||
AK312597.1 | - | 14 | 1798 | |||
AK316031.1 | - | 4 | 611 | |||
BC013903.2 | - | 13 | 1805 | 2959399 | ||
BC068458.1 | - | 13 | 1814 | 4819793 | ||
FJ224318.1 | - | 13 | 1790 | |||
J05021.1 | - | 13 | 1749 | |||
X51521.1 | - | 13 | 1790 |