Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003380.3 | NP_003371.2 | 8, 9 | 1692 | ||
XM_006717500.1 | XP_006717563.1 | 7, 8 | 2767 | |||
GenBank mRNA | AF328728.1 | - | 7, 8 | 1408 | ||
AK091813.1 | - | 8, 9 | 1383 | |||
AK097336.1 | - | 8, 9 | 1388 | |||
AK098444.1 | - | 8, 9 | 1262 | |||
AK222482.1 | - | 7, 8 | 1422 | |||
AK222507.1 | - | 7, 8 | 1424 | |||
AK222602.1 | - | 7, 8 | 1424 | |||
AK290643.1 | - | 7, 8 | 1304 | |||
BC000163.2 | - | 7, 8 | 1403 | 2985712 | ||
BC030573.2 | - | 7, 8 | 1403 | 3637336 | ||
BC066956.1 | - | 8, 9 | 1480 | 4823475 | ||
CR407690.1 | - | 7, 8 | 1279 | |||
EU794672.1 | - | 7, 8 | 1424 | |||
KU178388.1 | - | 7, 8 | 1279 | |||
KU178389.1 | - | 8, 9 | 896 | |||
M25246.1 | - | 7, 8 | 783 | |||
X56134.1 | - | 7, 8 | 1322 | |||
Z19554.2 | - | 7, 8 | 1401 |