Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF133 SIRNA Assays ›
Gene Symbol:
ZNF133
Gene Name:
zinc finger protein 133
Gene Aliases:
ZNF150, pHZ-13, pHZ-66
Chromosome Location:
Chr.20: 18288228 - 18316996 on Build GRCh38
UniGene ID:
Hs.472221
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139040
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF133
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083330.2
NP_001076799.1
4
2285
NM_001282995.1
NP_001269924.1
6
2521
NM_001282996.1
NP_001269925.1
6
2395
NM_001282997.1
NP_001269926.1
5
2446
NM_001282998.1
NP_001269927.1
4
2275
NM_001282999.1
NP_001269928.1
4
2288
NM_001283000.1
NP_001269929.1
5
2464
NM_001283001.1
NP_001269930.1
5
2451
NM_001283002.1
NP_001269931.1
5
2353
NM_001283003.1
NP_001269932.1
5
2426
NM_001283004.1
NP_001269933.1
5
2413
NM_001283005.1
NP_001269934.1
2
2049
NM_001283006.1
NP_001269935.1
3
2127
NM_001283007.1
NP_001269936.1
3
2145
NM_001283008.1
NP_001269937.1
4
2315
NM_003434.5
NP_003425.2
7
2697
XM_005260819.1
XP_005260876.1
4
2097
XM_005260820.4
XP_005260877.1
6
2354
XM_011529336.1
XP_011527638.1
7
2433
XM_011529337.1
XP_011527639.1
6
2305
XM_011529338.2
XP_011527640.1
6
2299
XM_011529339.1
XP_011527641.1
7
2433
XM_011529340.1
XP_011527642.1
2
1908
XM_017028041.1
XP_016883530.1
5
10198
XM_017028042.1
XP_016883531.1
5
3472
XM_017028043.1
XP_016883532.1
5
10195
XM_017028044.1
XP_016883533.1
7
10545
XM_017028045.1
XP_016883534.1
5
10613
XM_017028046.1
XP_016883535.1
5
10211
XM_017028047.1
XP_016883536.1
5
3565
XM_017028048.1
XP_016883537.1
4
3394
XM_017028049.1
XP_016883538.1
7
10555
XM_017028050.1
XP_016883539.1
7
2453
XM_017028051.1
XP_016883540.1
8
2629
XM_017028052.1
XP_016883541.1
6
2293
XM_017028053.1
XP_016883542.1
4
2327
XM_017028054.1
XP_016883543.1
5
2207
XM_017028055.1
XP_016883544.1
8
2623
XM_017028056.1
XP_016883545.1
10
2842
XM_017028057.1
XP_016883546.1
7
2410
XM_017028058.1
XP_016883547.1
8
2627
GenBank mRNA
AK123005.1
-
2
3863
AK291389.1
-
5
2203
AK294263.1
-
3
1876
AK295273.1
-
5
2125
AK295513.1
-
5
2189
AK313525.1
-
4
2024
AK316386.1
-
2
1803
BC001887.1
-
7
2503
3535533
BT007310.1
-
3
1944
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline