Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZYX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010972.1 | NP_001010972.1 | 10 | 2033 | ||
NM_003461.4 | NP_003452.1 | 10 | 2036 | |||
XM_011516569.2 | XP_011514871.2 | 9 | 1931 | |||
XM_017012587.1 | XP_016868076.1 | 10 | 2126 | |||
GenBank mRNA | AK130138.1 | - | 6 | 1326 | ||
AK298925.1 | - | 8 | 1782 | |||
AK299005.1 | - | 9 | 1752 | |||
BC002323.2 | - | 10 | 1905 | |||
BC008743.2 | - | 10 | 2000 | 3345843 | ||
BC009360.2 | - | 10 | 1945 | 4301542 | ||
BC010031.2 | - | 10 | 1933 | 3506023 | ||
BC017183.2 | - | 9 | 1562 | |||
U15158.1 | - | 8 | 1641 | |||
X94991.1 | - | 9 | 1888 | |||
X95735.1 | - | 10 | 1949 |