Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003468.3 | NP_003459.2 | 2 | 2366 | ||
GenBank mRNA | AB043702.1 | - | 2 | 2341 | ||
U43318.1 | - | 2 | 2276 |
Set Membership: |
G-protein coupled receptor |