Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078645.1 | NP_001072113.1 | 5 | 470 | ||
NM_001318517.1 | NP_001305446.1 | 5 | 467 | |||
NM_001318518.1 | NP_001305447.1 | 5 | 467 | |||
NM_003903.3 | NP_003894.3 | 5 | 470 | |||
XM_005266206.1 | XP_005266263.1 | 5 | 512 | |||
XM_006719992.1 | XP_006720055.1 | 5 | 457 | |||
XM_006719993.1 | XP_006720056.1 | 4 | 405 | |||
XM_011534851.1 | XP_011533153.1 | 5 | 512 | |||
XM_011534853.1 | XP_011533155.1 | 5 | 512 | |||
XM_017020830.1 | XP_016876319.1 | 5 | 512 | |||
XM_017020831.1 | XP_016876320.1 | 5 | 512 | |||
XM_017020832.1 | XP_016876321.1 | 4 | 405 | |||
XM_017020833.1 | XP_016876322.1 | 3 | 90 | |||
GenBank mRNA | AB209850.1 | - | 4 | 1623 | ||
AF164598.1 | - | 5 | 273 | |||
AK296424.1 | - | 5 | 460 | |||
AK315302.1 | - | 5 | 301 | |||
AL540490.3 | - | 5 | 466 | |||
BC010875.2 | - | 5 | 414 | 3879150 | ||
BC017244.1 | - | 5 | 380 | 4877392 | ||
DC323610.1 | - | 5 | 473 | |||
U18291.1 | - | 5 | 293 |