Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135774.1 | NP_001129246.1 | 14 | 1994 | ||
NM_003490.3 | NP_003481.3 | 13 | 1894 | |||
NM_133633.2 | NP_598344.2 | 12 | 1602 | |||
XM_011530405.2 | XP_011528707.1 | 14 | 2530 | |||
XM_011530406.2 | XP_011528708.1 | 14 | 1996 | |||
XM_011530407.2 | XP_011528709.1 | 14 | 2403 | |||
XM_011530408.1 | XP_011528710.1 | 14 | 1994 | |||
XM_011530410.2 | XP_011528712.1 | 12 | 2198 | |||
XM_017028960.1 | XP_016884449.1 | 15 | 2928 | |||
XM_017028961.1 | XP_016884450.1 | 14 | 2320 | |||
XM_017028962.1 | XP_016884451.1 | 14 | 2255 | |||
XM_017028963.1 | XP_016884452.1 | 14 | 1919 | |||
XM_017028964.1 | XP_016884453.1 | 14 | 1991 | |||
XM_017028965.1 | XP_016884454.1 | 8 | 1765 | |||
GenBank mRNA | AB209684.1 | - | 14 | 1946 | ||
AF046873.1 | - | 13 | 1771 | |||
AK300171.1 | - | 6 | 727 | |||
BC075065.2 | - | 13 | 1761 | 30915588 | ||
BC075066.2 | - | 13 | 1761 | 30915375 | ||
BC117460.1 | - | 13 | 1759 | 40126011 | ||
BC143874.1 | - | 13 | 1762 | 9052390 | ||
CR456589.1 | - | 13 | 1826 |