Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003551.2 | NP_003542.1 | 4 | 427 | ||
XM_005272099.1 | XP_005272156.1 | 3 | 354 | |||
XM_006714716.3 | XP_006714779.1 | 3 | 354 | |||
XM_017009945.1 | XP_016865434.1 | 3 | 354 | |||
GenBank mRNA | AF067724.1 | - | 4 | 397 | ||
AK312181.1 | - | 4 | 374 | |||
BC026182.2 | - | 4 | 399 | 5270050 | ||
KU178416.1 | - | 3 | 349 | |||
U90450.1 | - | 4 | 373 | |||
Y14992.1 | - | 3 | 363 |