Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007226.1 | NP_001007227.1 | 8 | 1115 | ||
NM_001007227.1 | NP_001007228.1 | 7 | 993 | |||
NM_001007228.1 | NP_001007229.1 | 6 | 941 | |||
NM_001007229.1 | NP_001007230.1 | 7 | 992 | |||
NM_001007230.1 | NP_001007231.1 | 8 | 1044 | |||
NM_003563.3 | NP_003554.1 | 7 | 1063 | |||
XM_005257723.4 | XP_005257780.1 | 7 | 1287 | |||
XM_005257724.4 | XP_005257781.1 | 6 | 1217 | |||
XM_017025204.1 | XP_016880693.1 | 5 | 730 | |||
GenBank mRNA | AJ000644.1 | - | 6 | 801 | ||
AK222589.1 | - | 6 | 766 | |||
AK293045.1 | - | 7 | 1274 | |||
AK293587.1 | - | 6 | 747 | |||
AK300616.1 | - | 6 | 765 | |||
AK303268.1 | - | 5 | 687 | |||
AK312691.1 | - | 6 | 777 | |||
AK316366.1 | - | 8 | 842 | |||
BC001269.1 | - | 7 | 947 | 3461565 | ||
BC003385.1 | - | 6 | 758 | 3445878 |