Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007226.1 | NP_001007227.1 | 12 | 1930 | ||
NM_001007227.1 | NP_001007228.1 | 11 | 1808 | |||
NM_001007228.1 | NP_001007229.1 | 10 | 1756 | |||
NM_001007229.1 | NP_001007230.1 | 11 | 1807 | |||
NM_001007230.1 | NP_001007231.1 | 12 | 1859 | |||
NM_003563.3 | NP_003554.1 | 11 | 1878 | |||
XM_005257723.4 | XP_005257780.1 | 11 | 2102 | |||
XM_005257724.4 | XP_005257781.1 | 10 | 2032 | |||
XM_017025204.1 | XP_016880693.1 | 9 | 1545 | |||
GenBank mRNA | AJ000644.1 | - | 10 | 1616 | ||
AK123385.1 | - | 1 | 1588 | |||
AK222589.1 | - | 10 | 1581 | |||
AK293587.1 | - | 10 | 1562 | |||
BC001269.1 | - | 11 | 1762 | 3461565 | ||
BC003385.1 | - | 10 | 1573 | 3445878 | ||
BM693397.1 | - | 1 | 284 | |||
W84567.1 | - | 1 | 235 |