Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003568.2 | NP_003559.2 | 13 | 1339 | ||
GenBank mRNA | AF230929.1 | - | 13 | 1095 | ||
AJ009985.1 | - | 13 | 1284 | |||
AK314926.1 | - | 12 | 1192 | |||
BC005830.2 | - | 12 | 995 | 2959488 | ||
CR536481.1 | - | 11 | 848 |