Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032296.3 | NP_001027467.2 | 7 | 1183 | ||
NM_001286649.1 | NP_001273578.1 | 6 | 1126 | |||
NM_003576.4 | NP_003567.2 | 7 | 1127 | |||
XM_005254079.4 | XP_005254136.1 | 7 | 1004 | |||
XM_017020794.1 | XP_016876283.1 | 7 | 1300 | |||
XM_017020795.1 | XP_016876284.1 | 7 | 962 | |||
GenBank mRNA | AF024636.1 | - | 7 | 896 | ||
AF083420.1 | - | 7 | 999 | |||
AF339826.1 | - | 6 | 604 | |||
AK297583.1 | - | 7 | 899 | |||
AK299140.1 | - | 6 | 843 | |||
BC035578.1 | - | 7 | 1131 | 5539375 | ||
BC065378.1 | - | 7 | 1127 | 6598075 | ||
BT019619.1 | - | 7 | 818 | |||
CR542227.1 | - | 7 | 818 | |||
EU668350.1 | - | 7 | 1174 |