Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008895.2 | NP_001008895.1 | 18 | 2037 | ||
NM_001278513.1 | NP_001265442.1 | 18 | 2181 | |||
NM_001278514.1 | NP_001265443.1 | 18 | 1849 | |||
NM_003589.2 | NP_003580.1 | 18 | 1863 | |||
XM_011537523.2 | XP_011535825.1 | 17 | 2397 | |||
GenBank mRNA | AB012193.1 | - | 12 | 1211 | ||
AB178950.1 | - | 18 | 1914 | |||
AF077188.1 | - | 18 | 1774 | |||
AK296700.1 | - | 10 | 1158 | |||
AK314835.1 | - | 18 | 2136 | |||
AL833355.1 | - | 17 | 1801 | |||
AY365124.1 | - | 18 | 1914 | |||
BC008308.1 | - | 18 | 1925 | 3537176 | ||
U58090.1 | - | 8 | 901 |