Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257197.1 | NP_001244126.1 | 2 | 520 | ||
NM_001257198.1 | NP_001244127.1 | 3 | 401 | |||
NM_003590.4 | NP_003581.1 | 3 | 718 | |||
XM_006712800.2 | XP_006712863.2 | 3 | 391 | |||
XM_011511995.1 | XP_011510297.1 | 3 | 303 | |||
XM_011511996.2 | XP_011510298.1 | 3 | 367 | |||
GenBank mRNA | AB014517.2 | - | 3 | 646 | ||
AF062537.1 | - | 3 | 419 | |||
AF064087.1 | - | 3 | 411 | |||
AK291151.1 | - | 3 | 368 | |||
AK299644.1 | - | 2 | 128 | |||
AK308496.1 | - | 3 | 401 | |||
AY337761.1 | - | 3 | 864 | |||
BC039598.1 | - | 3 | 663 | 5784147 | ||
BC092409.1 | - | 3 | 663 | 6168545 |